Learn More
Biocare Medical, LLC TTF-1 + Napsin A, 6ml, Provides a High Sensitivity and Specificity for Lung Adenocarcinoma
SDP
Catalog No. 5083229
Encompass_Preferred
TTF-1 + Napsin A provides a high sensitivity and specificity for lung adenocarcinomas. When used in a panel with p63 and CK5, it may aid in the analysis of poorly differentiated lung adenocarcinomas vs. squamous cell carcinomas.
Catalog No. 50-832-29 Supplier Biocare Medical, LLC Supplier No. PPM394DSAA
TTF-1 + Napsin A provides a high sensitivity and specificity for lung adenocarcinomas. When used in a panel with p63 and CK5, it may aid in the analysis of poorly differentiated lung adenocarcinomas vs. squamous cell carcinomas.