Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7080MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: NX2.1/690. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: MAD109, MLE-15, H441-4, or H345 cells. Normal thyroid or lung. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant TTF-1 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 40kDa.Chromosome: 14q13.3. Gene Symbol: NKX2-1. Gene ID: 7080. Ensemble: ENSG00000136352. Uniprot: NP_003308.
Catalog No. 50-188-5226