Learn More
Aviva Systems Biology Corp Ttf1 Antibody - N-terminal region (ARP36997_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36997_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ttf1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of mouse Ttf1. Alternate names: AV245725ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-561