Learn More
Aviva Systems Biology Corp TTF1 Antibody - middle region (ARP61242_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61242_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TTF1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TTF1. Alternate names: TTF-1, TTF-IApplicationWB. Species reactivity: Human
Catalog No. 50-287-311