Learn More
INTACT GENOMICS INC Tth DNA Ligase, 1000 units
Catalog No. NC3998109
Encompass
High-fidelity Thermus thermophilus (Tth) DNA Ligase from Intact Genomics delivers precise, thermostable ligation with excellent SNP discrimination and low background—ideal for LCR and mutation detection.
Catalog No. NC3998109 Supplier INTACT GENOMICS INC Supplier No. 3223
High-fidelity Thermus thermophilus (Tth) DNA Ligase from Intact Genomics delivers precise, thermostable ligation with excellent SNP discrimination and low background—ideal for LCR and mutation detection.