Learn More
New England Biolabs, Inc. Tth Endonuclease IV – 500 units
SDP
Catalog No. 501526388
Encompass
Tth Endonuclease IV is a thermostable apurinic/apyrimidinic (AP) endonuclease from Thermus thermophilus.
Catalog No. 50-152-6388 Supplier New England Biolabs, Inc. Supplier No. M0294S
Tth Endonuclease IV is a thermostable apurinic/apyrimidinic (AP) endonuclease from Thermus thermophilus.