Learn More
New England Biolabs, Inc. Tth111I – 400 units
SDP
Catalog No. 50812103
Encompass
A restriction endonuclease that recognizes the sequence GACN^N_NGTC.
Catalog No. 50-812-103 Supplier New England Biolabs, Inc. Supplier No. R0185S
A restriction endonuclease that recognizes the sequence GACN^N_NGTC.