Learn More
Aviva Systems Biology Corp TTL Antibody - middle region (ARP53199_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53199_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TTL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TTL. Alternate names: MGC46235ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60967