Learn More
Crystal Chem Inc Total Testosterone ELISA Kit (Saliva), 96-wells
SDP
Supplier:  Crystal Chem Inc 80981
Encompass
The Total Testosterone ELISA (Saliva) is used to quantify levels of human total testosterone in saliva using only a 100 µL sample volume. Highlights: Kits use sample size of only 100 µL. High sensitivity of 1 pg/mL. Dynamic range of 2 - 800 pg/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7882