Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metro TTM18C Top-Track Mobile Unit Kit for 18 in. Wide Super Erecta Wire Shelving

Catalog No. 19138219 Shop All Metro Products
Change view
Click to view available options
For Use With (Equipment):
Metro Top-Track
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-138-219 Metro Top-Track
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-138-219 Supplier Metro Supplier No. TTM18C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maximize your storage capacity with Top-Track overhead track shelving. Top-Track system allow more storage units to fit in a defined space than a traditional shelving setup.

  • A movable, open aisle allows full access to each entire shelving unit thus eliminating hard to reach or dead spaces.
  • Includes four 74 in. (1880 mm) high mobile posts, four plated casters, donut bumpers, guide rollers, and other hardware to assemble a mobile unit.
  • Shelves are sold separately.
  • Maximum allowable shelf length is 60 in. (1524 mm) when using 18 in. (457 mm) wide mobile units.
  • Maximum of five 18 in. (457 mm) units are allowable between two stationary units.

Specifications

Product Type Mobile Shelving Post
Color Silver
Material Steel
For Use With (Application) Save Space with High Density storage
For Use With (Equipment) Metro Top-Track
Includes Mobile Unit for Metro Top-Track
Width (English) 18 in.
Width (Metric) 457.2 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.