Learn More
TARGETMOL CHEMICALS INC TTNPB 25MG
SDP
Catalog No. 502581407
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TTNPB (Ro 13-7410AGN-191183) a potent RAR agonist inhibits binding of [3H]tRA of human RAR(alpha) (IC50 5.1 nM) (beta) (IC50 4.5 nM) and (gamma) (IC50 9.3 nM) respectively. purity: 98%
Catalog No. 50-258-1407 Supplier TARGETMOL CHEMICALS INC Supplier No. T128825MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TTNPB (Ro 13-7410AGN-191183) a potent RAR agonist inhibits binding of [3H]tRA of human RAR(alpha) (IC50 5.1 nM) (beta) (IC50 4.5 nM) and (gamma) (IC50 9.3 nM) respectively. purity: 98%