Learn More
Enzo Life Sciences TTNPB (5mg). CAS: 71441-28-6
SDP
Supplier:  Enzo Life Sciences BMLGR1050005
Encompass_Preferred
Retinoic acid analog which acts as a selective RAR agonist. Alternative name: Ro 13-7410, 4-[E-2-(5,6,7,8-Tetrahydro-5,5,8,8-tetra-methyl-2-naphthalenyl)-1-propenyl]benzoic acid. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO or ethanol (with gentle warming). Long Term Storage: -20°C.
Catalog No. 50-201-0918