Learn More
Selleck Chemical LLC TTNPB (Arotinoid Acid) 10mg 71441-28-6 Ro 13-7410, AGN-191183
SDP
Supplier:  Selleck Chemical LLC S462710MG
Encompass_Preferred
TTNPB (Arotinoid Acid, Ro 13-7410, AGN-191183) is a potent RAR agonist, and inhibits binding of [3H]tRA with IC50 of 5.1 nM, 4.5 nM, and 9.3 nM for human RAR?, ?, and ?, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4076