Learn More
American Radiolabeled Chemicals Inc TTP 1,2-3H 1LC
SDP
Catalog No. 50353062
Encompass
Thymidine 5'-methyl-1',2'-3H triphosphate ammonium salt (TTP) 1mCi in ethanol:water (1:1), 1 mCi/ml, 80-120 Ci/mmol, M.W. 501.21, Shipped in Dry Ice, Exclusive
Catalog No. 50-353-062 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 19301MC
Thymidine 5'-methyl-1',2'-3H triphosphate ammonium salt (TTP) 1mCi in ethanol:water (1:1), 1 mCi/ml, 80-120 Ci/mmol, M.W. 501.21, Shipped in Dry Ice, Exclusive