Learn More
Midwest Scientific 12-well TC Plate, flat bottom, 126/cs
SDP
Catalog No. NC1848593
Encompass
Air vents in the base of the plates allow for consistent air flow between stacked plates. The consistent air flow ensures even temperature distribution, condensation prevention, and greater well to well consistency.
Catalog No. NC1848593 Supplier Midwest Scientific Supplier No. TP92012
Add to Cart
Add to Cart
Air vents in the base of the plates allow for consistent air flow between stacked plates. The consistent air flow ensures even temperature distribution, condensation prevention, and greater well to well consistency.