Learn More
Antibodies Online TTR ELISA Kit, Sandwich enzyme immunoassay, 96-well plate format, Detection range: 0.31 ng/mL - 20 ng/mL, Sensitivity: 0.151 ng/mL, Reactive with human samples
SDP
Supplier:  Antibodies Online ABIN6574196
Encompass
TTR ELISA Kit, Sandwich enzyme immunoassay, 96-well plate format, Detection range: 0.31 ng/mL - 20 ng/mL, Sensitivity: 0.151 ng/mL, Reactive with human samples
Catalog No. NC2306769