Learn More
Bioworld TTTRIS-TRICINE) BUFFER 1
Catalog No. 50198656
Encompass_Preferred
TT (TRIS-TRICINE) BUFFER, 10X SOLUTION: Tris-Tricine Tris-Tricine (TT) Buffer has been developed for both non-denaturing and denaturing electrophoresis of proteins. TT Buffer offers several advantages over more conventionally used protei...
Catalog No. 50-198-656 Supplier Bioworld Supplier No. 214200251
TT (TRIS-TRICINE) BUFFER, 10X SOLUTION: Tris-Tricine Tris-Tricine (TT) Buffer has been developed for both non-denaturing and denaturing electrophoresis of proteins. TT Buffer offers several advantages over more conventionally used protei...