Learn More
ERA WATERS CORP TT USP System Suitability Set, 1L
SDP
Catalog No. NC1792860
Encompass
Each system suitability set contains (1) Reagent Water Rw, (1) 0.500 mg/L C from USP Sucrose Rs and (1) 0.500 mg/L C from USP 1,4-benzoquinone Rss.
Catalog No. NC1792860 Supplier ERA WATERS CORP Supplier No. 18204
Each system suitability set contains (1) Reagent Water Rw, (1) 0.500 mg/L C from USP Sucrose Rs and (1) 0.500 mg/L C from USP 1,4-benzoquinone Rss.