Learn More
Aviva Systems Biology Corp TTYH3 Antibody - middle region (ARP49946_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49946_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TTYH3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TTYH3. Alternate names: KIAA1691ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-466