Learn More
Azenta US Inc TU,0.9,EXT,2CD,UCP,HBRK,96RK,1
Catalog No. NC2079331
Encompass
TU,0.9,EXT,2CD,UCP,HBRK,96RK,10CA0.9ml Dual-coded Tube, 96-format, External Thread, 2D Codeand Human Readable Number on base, uncapped, 10 racksper case, HighBase rack, empty rack part number: 66-61002
This item is not returnable. View return policy
Catalog No. NC2079331 Supplier Azenta US Inc Supplier No. 68100101
TU,0.9,EXT,2CD,UCP,HBRK,96RK,10CA0.9ml Dual-coded Tube, 96-format, External Thread, 2D Codeand Human Readable Number on base, uncapped, 10 racksper case, HighBase rack, empty rack part number: 66-61002