Learn More
Hach Company Hach TU5 Series TU5200 Laboratory Laser Turbidimeter with RFID, EPA Version
Catalog No. NC1942018 Shop All Hach Company Products
Encompass
The Hach TU5 Series TU5200 laboratory laser turbidimeter with RFID (EPA version) is a benchtop instrument for drinking water and other pure water testing. It combines advanced detection optics with a color touchscreen and RFID sample tagging to deliver fast, repeatable, EPA-compliant turbidity measurements.
  • Provides measurement ranges up to 700 NTU with alternative units for FNU, FTU, mg/L, and EBC.
  • 360° x 90° detection technology reduces sample surface area cleaning requirements.
  • RFID sample tagging stores and transfers sample information between compatible instruments.
  • 7 in color WVGA touchscreen with intuitive, touch-based operation.
  • High repeatability across the measurement range for reliable results.
  • Compact benchtop footprint and lightweight enclosure suitable for laboratory use.
  • Wide power input (100-240 VAC, 50/60 Hz) for global compatibility.
Catalog No. NC1942018 Supplier Hach Company Supplier No. LPV442.99.03012
Add to Cart
Add to Cart
Request a Quote
The Hach TU5 Series TU5200 laboratory laser turbidimeter with RFID (EPA version) is a benchtop instrument for drinking water and other pure water testing. It combines advanced detection optics with a color touchscreen and RFID sample tagging to deliver fast, repeatable, EPA-compliant turbidity measurements.
  • Provides measurement ranges up to 700 NTU with alternative units for FNU, FTU, mg/L, and EBC.
  • 360° x 90° detection technology reduces sample surface area cleaning requirements.
  • RFID sample tagging stores and transfers sample information between compatible instruments.
  • 7 in color WVGA touchscreen with intuitive, touch-based operation.
  • High repeatability across the measurement range for reliable results.
  • Compact benchtop footprint and lightweight enclosure suitable for laboratory use.
  • Wide power input (100-240 VAC, 50/60 Hz) for global compatibility.