Learn More
Hach Company TU5200 Turb w/o RFID ISO Ver.
Catalog No. 5000208133 Shop All Hach Company Products
Encompass
TU5200 benchtop turbidimeter kit includes user manual-Stablcal kit (10-20 and 600 NTU)-2 sample vials-power supply-dust cover-vial stand
Catalog No. 50-002-08133 Supplier Hach Company Supplier No. LPV442.99.01022
Add to Cart
Add to Cart
Request a Quote
TU5200 benchtop turbidimeter kit includes user manual-Stablcal kit (10-20 and 600 NTU)-2 sample vials-power supply-dust cover-vial stand