Learn More
Hach Company Hach TU5300sc TU5 Series Low Range Laser Turbidimeter with Flow Sensor, Automatic Cleaning, RFID, System Check (EPA version)
Catalog No. NC2173622 Shop All Hach Company Products
Encompass
The TU5300sc low range laser turbidimeter provides high-sensitivity turbidity measurement for potable and high-purity water applications. Its wide-angle nephelometric optics and integrated maintenance features deliver reliable, repeatable readings with reduced manual maintenance.
  • Measurement range: 0-700 NTU (EPA version).
  • Accuracy: ±2% or 0.01 NTU from 0-40 NTU; ±10% from 40-1000 NTU (Formazin, 25 °C).
  • Sample flow rate: 100-1,000 mL/min (optimal 200-500 mL/min).
  • Includes automatic cleaning module and external flow sensor.
  • Integrated RFID and System Check for diagnostics and maintenance tracking.
  • Requires an external digital controller for operation; power supplied by controller.
  • Compact enclosure: 249 x 268 x 190 mm; approx. 2.7 kg (5.95 lb) without accessories.
Catalog No. NC2173622 Supplier Hach Company Supplier No. LXV445.99.53112
The TU5300sc low range laser turbidimeter provides high-sensitivity turbidity measurement for potable and high-purity water applications. Its wide-angle nephelometric optics and integrated maintenance features deliver reliable, repeatable readings with reduced manual maintenance.
  • Measurement range: 0-700 NTU (EPA version).
  • Accuracy: ±2% or 0.01 NTU from 0-40 NTU; ±10% from 40-1000 NTU (Formazin, 25 °C).
  • Sample flow rate: 100-1,000 mL/min (optimal 200-500 mL/min).
  • Includes automatic cleaning module and external flow sensor.
  • Integrated RFID and System Check for diagnostics and maintenance tracking.
  • Requires an external digital controller for operation; power supplied by controller.
  • Compact enclosure: 249 x 268 x 190 mm; approx. 2.7 kg (5.95 lb) without accessories.