Learn More
Hach Company TU5300sc LR AutoClean RFID
Catalog No. 5000207962 Shop All Hach Company Products
Encompass
TU5 Series TU5300sc Low Range Laser Turbidimeter with Automatic Cleaning and RFID-EPA VersionIncludesTU5300sc Turbidimeter-Automatic Cleaning Unit-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket
Catalog No. 50-002-07962 Supplier Hach Company Supplier No. LXV445.99.32112
TU5 Series TU5300sc Low Range Laser Turbidimeter with Automatic Cleaning and RFID-EPA VersionIncludesTU5300sc Turbidimeter-Automatic Cleaning Unit-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket