Learn More
Hach Company TU5300sc LR Laser Turb-ISO
Catalog No. 5000207491 Shop All Hach Company Products
Encompass
TU5 Series TU5300sc Low Range Laser Turbidimeter-ISO VersionIncludesTU5300sc Turbidimeter-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket
Catalog No. 50-002-07491 Supplier Hach Company Supplier No. LXV445.99.10122
TU5 Series TU5300sc Low Range Laser Turbidimeter-ISO VersionIncludesTU5300sc Turbidimeter-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket