Learn More
Hach Company TU5300sc LR Turb AutoClean EPA
Catalog No. 5000208073 Shop All Hach Company Products
Encompass
TU5 Series TU5300sc Low Range Laser Turbidimeter with Automatic Cleaning-EPA VersionIncludesTU5300sc Turbidimeter-Automatic Cleaning Unit-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket
Catalog No. 50-002-08073 Supplier Hach Company Supplier No. LXV445.99.30112
TU5 Series TU5300sc Low Range Laser Turbidimeter with Automatic Cleaning-EPA VersionIncludesTU5300sc Turbidimeter-Automatic Cleaning Unit-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket