Learn More
Hach Company TU5300sc LR Turb Flow RFID EPA
Catalog No. 5000208053 Shop All Hach Company Products
Encompass
TU5 Series TU5300sc Low Range Laser Turbidimeter with Flow Sensor and RFID-EPA VersionIncludesTU5300sc Turbidimeter-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket-Flow Sensor
Catalog No. 50-002-08053 Supplier Hach Company Supplier No. LXV445.99.22112
TU5 Series TU5300sc Low Range Laser Turbidimeter with Flow Sensor and RFID-EPA VersionIncludesTU5300sc Turbidimeter-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket-Flow Sensor