Learn More
Hach Company TU5300sc LR Turb w/ChkRFID ISO
Catalog No. 5000208070 Shop All Hach Company Products
Encompass
TU5 Series TU5300sc Low Range Laser Turbidimeter with System Check and RFID-ISO VersionIncludesTU5300sc Turbidimeter-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket
Catalog No. 50-002-08070 Supplier Hach Company Supplier No. LXV445.99.13122
TU5 Series TU5300sc Low Range Laser Turbidimeter with System Check and RFID-ISO VersionIncludesTU5300sc Turbidimeter-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket