Learn More
Hach Company TU5300sc TurbEPA w/SC4500 1Ch
Supplier:  Hach Company LXV525.97KTO012
Encompass
TU5300sc Low Range Laser Turbidimeter-EPA Version-Automatic Cleaning Module Flow with SC4500 Controller-1 ChannelIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07360
Add to Cart
Request a Quote