Learn More
Hach Company TU5300sc TurbEPA w/SC4500 1Ch
Catalog No. 5000208061 Shop All Hach Company Products
Encompass
TU5300sc Low Range Laser Turbidimeter-EPA-Automatic Cleaning Module-System Check-RFID-Flow-w/ SC4500 Controller-1 ChannelIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-08061 Supplier Hach Company Supplier No. LXV525.97KTO007
Add to Cart
Add to Cart
Request a Quote
TU5300sc Low Range Laser Turbidimeter-EPA-Automatic Cleaning Module-System Check-RFID-Flow-w/ SC4500 Controller-1 ChannelIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware