Learn More
Hach Company TU5300sc TurbISOw/SC4500 1Ch
Catalog No. 5000207970 Shop All Hach Company Products
Encompass
TU5300sc Low Range Laser Turbidimeter ISO Version System Check RFID Flow with SC4500 Controller-1 ChannelIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07970 Supplier Hach Company Supplier No. LXV525.97KTO009
Add to Cart
Add to Cart
Request a Quote
TU5300sc Low Range Laser Turbidimeter ISO Version System Check RFID Flow with SC4500 Controller-1 ChannelIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware