Learn More
Hach Company TU5300scLR Turb w/SC4500
Catalog No. 5000207926 Shop All Hach Company Products
Encompass
TU5300sc Low Range Laser Turbidimeter-ISO Version System Check RFID Flow with SC4500 Controller-2 ChannelsIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07926 Supplier Hach Company Supplier No. LXV525.97KTO043
Add to Cart
Add to Cart
Request a Quote
TU5300sc Low Range Laser Turbidimeter-ISO Version System Check RFID Flow with SC4500 Controller-2 ChannelsIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware