Learn More
Hach Company TU5300scLR Turb w/SC4500
Catalog No. 5000207975 Shop All Hach Company Products
Encompass
TU5300sc Low Range Laser Turbidimeter-ISO-Auto Cleaning-System Check-RFID-Flow-SC4500 Controller-2 ChannelsIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07975 Supplier Hach Company Supplier No. LXV525.97KTO042
Add to Cart
Add to Cart
Request a Quote
TU5300sc Low Range Laser Turbidimeter-ISO-Auto Cleaning-System Check-RFID-Flow-SC4500 Controller-2 ChannelsIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware