Learn More
Hach Company TU5400 sc Panel
Supplier:  Hach Company SPMPTU5400
Encompass
The single parameter turbidity panels offers the TU5400sc Ultra-High precision Low Range Laser Turbidimeter and SC4500 controller in the convenience of one panel These panel configurations are pre-engineered ready to install simple solutions that are designed to work together IncludesTU5400sc Turbidimeter (LXV445 53 B3212)SC4500 Controller-Claros-enabled-5x mA Output-2 digital Sensors-100-240 VAC-US plug (LXV525 99EA1551)Flow Sensor for TU5300sc and TU5400sc Laser Turbidimeter (LQV160 99 00002)
Catalog No. 50-002-08040
Add to Cart
Request a Quote