Learn More
Hach Company TU5400sc Prec LR AutoClean Chk
Catalog No. 5000207797 Shop All Hach Company Products
Encompass
TU5 Series TU5400sc Ultra-High Precision Low Range Laser Turbidimeter with Automatic Cleaning and System Check-ISO VersionIncludesTU5400sc Turbidimeter-Automatic Cleaning Unit-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket
Catalog No. 50-002-07797 Supplier Hach Company Supplier No. LXV445.99.31222
Add to Cart
Add to Cart
Request a Quote
TU5 Series TU5400sc Ultra-High Precision Low Range Laser Turbidimeter with Automatic Cleaning and System Check-ISO VersionIncludesTU5400sc Turbidimeter-Automatic Cleaning Unit-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket