Learn More
Hach Company TU5400sc Prec LR AutoClean ISO
Catalog No. 5000208028 Shop All Hach Company Products
Encompass
TU5 Series TU5400sc Ultra-High Precision Low Range Laser Turbidimeter with Automatic Cleaning-ISO VersionIncludesTU5400sc Turbidimeter-Automatic Cleaning Unit-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket
Catalog No. 50-002-08028 Supplier Hach Company Supplier No. LXV445.99.30222
Add to Cart
Add to Cart
Request a Quote
TU5 Series TU5400sc Ultra-High Precision Low Range Laser Turbidimeter with Automatic Cleaning-ISO VersionIncludesTU5400sc Turbidimeter-Automatic Cleaning Unit-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket