Learn More
Hach Company TU5400sc Prec LR Turb FlowRFID
Catalog No. 5000208013 Shop All Hach Company Products
Encompass
TU5 Series TU5400sc Ultra-High Precision Low Range Laser Turbidimeter with Flow Sensor and RFID-EPA VersionIncludesTU5400sc Turbidimeter-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket-Flow Sensor
Catalog No. 50-002-08013 Supplier Hach Company Supplier No. LXV445.99.22212
Add to Cart
Add to Cart
Request a Quote
TU5 Series TU5400sc Ultra-High Precision Low Range Laser Turbidimeter with Flow Sensor and RFID-EPA VersionIncludesTU5400sc Turbidimeter-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket-Flow Sensor