Learn More
Hach Company TU5400sc Prec LR Turb w/RFID
Catalog No. 5000207954 Shop All Hach Company Products
Encompass
TU5 Series TU5400sc Ultra-High Precision Low Range Laser Turbidimeter with RFID-ISO VersionIncludesTU5400sc Turbidimeter-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket
Catalog No. 50-002-07954 Supplier Hach Company Supplier No. LXV445.99.12222
Add to Cart
Add to Cart
Request a Quote
TU5 Series TU5400sc Ultra-High Precision Low Range Laser Turbidimeter with RFID-ISO VersionIncludesTU5400sc Turbidimeter-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket