Learn More
Hach Company TU5400sc Precision w/SC4501
Catalog No. 5000207925 Shop All Hach Company Products
Encompass
TU5400sc Ultra-High Precision Low Range Laser Turbidimeter-EPA-Auto Cleaning-System Check-RFID-Flow-SC4500-2 ChannelsIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware
Catalog No. 50-002-07925 Supplier Hach Company Supplier No. LXV525.97KTO039
Add to Cart
Add to Cart
Request a Quote
TU5400sc Ultra-High Precision Low Range Laser Turbidimeter-EPA-Auto Cleaning-System Check-RFID-Flow-SC4500-2 ChannelsIncludes Controller-Non-Claros Embedded Prognosys-without power cord includes mounting hardware