Learn More
Hach Company TU5400sc Ultra Precision Turb
Catalog No. 5000208022 Shop All Hach Company Products
Encompass
TU5 Series TU5400sc Ultra-High Precision Low Range Laser Turbidimeter-Flow Sensor-Automatic Cleaning-RFID-System Check-ISOTU5400sc Turbidimeter-Automatic Cleaning Unit-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket-Flow Sensor
Catalog No. 50-002-08022 Supplier Hach Company Supplier No. LXV445.99.53222
Add to Cart
Add to Cart
Request a Quote
TU5 Series TU5400sc Ultra-High Precision Low Range Laser Turbidimeter-Flow Sensor-Automatic Cleaning-RFID-System Check-ISOTU5400sc Turbidimeter-Automatic Cleaning Unit-User Manual-Wall Mount-Vial Replacement Tool-Screw Set-Drying Cartridge-Flow Regulator-Service Bracket-Flow Sensor