Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, 1/4 in I.D., 50 ft, Textile Reinforced
Catalog No. 502365839
Encompass_Preferred
Saint-Gobain Tygon 2475 I.B. high-pressure thermoplastic tubing, 1/4 in I.D. x 1/2 in O.D., 50 ft pack, combines textile-braid reinforcement and a smooth, low-sorption inner surface for reliable, high-purity fluid transfer. Formulated without added plasticizers and with clear construction, it supports applications requiring low extractables, visual inspection, and elevated working pressures.
  • Textile-braid reinforcement for elevated working pressures.
  • No added plasticizers to minimize extractables.
  • Low-wetting, ultra-smooth inner surface to reduce sorption and microbial buildup.
  • Clear construction allows visual inspection of fluid path.
  • Dimensions: 1/4 in I.D., 1/2 in O.D., 50 ft pack length.
  • Lower gas permeation than standard silicone tubing.
Catalog No. 50-236-5839 Supplier Accuflow Systems Supplier No. ACX00019
Saint-Gobain Tygon 2475 I.B. high-pressure thermoplastic tubing, 1/4 in I.D. x 1/2 in O.D., 50 ft pack, combines textile-braid reinforcement and a smooth, low-sorption inner surface for reliable, high-purity fluid transfer. Formulated without added plasticizers and with clear construction, it supports applications requiring low extractables, visual inspection, and elevated working pressures.
  • Textile-braid reinforcement for elevated working pressures.
  • No added plasticizers to minimize extractables.
  • Low-wetting, ultra-smooth inner surface to reduce sorption and microbial buildup.
  • Clear construction allows visual inspection of fluid path.
  • Dimensions: 1/4 in I.D., 1/2 in O.D., 50 ft pack length.
  • Lower gas permeation than standard silicone tubing.