Learn More
Accuflow Systems Saint-Gobain Silicone Tubing, 1/16 in ID, 3/16 in OD
Catalog No. NC2323796
Encompass_Preferred
Saint-Gobain Tygon SPT-3350 sanitary silicone tubing is a food- and beverage-grade, ultra-smooth silicone tubing engineered to minimize particle entrapment and reduce extractables. This 1/16 in I.D. x 3/16 in O.D. configuration is supplied in 50 ft coils and provides excellent fluid flow for hygienic dispensing and transfer applications.
  • Sanitary, ultra-smooth inner bore reduces particle entrapment.
  • Food and beverage grade, FDA and NSF-51 compliant.
  • Minimal extractables preserve fluid integrity.
  • Size: 1/16 in I.D., 3/16 in O.D., 50 ft coil.
  • Flexible with excellent fluid flow and easy to clean.
Catalog No. NC2323796 Supplier Accuflow Systems Supplier No. ABW00003
Saint-Gobain Tygon SPT-3350 sanitary silicone tubing is a food- and beverage-grade, ultra-smooth silicone tubing engineered to minimize particle entrapment and reduce extractables. This 1/16 in I.D. x 3/16 in O.D. configuration is supplied in 50 ft coils and provides excellent fluid flow for hygienic dispensing and transfer applications.
  • Sanitary, ultra-smooth inner bore reduces particle entrapment.
  • Food and beverage grade, FDA and NSF-51 compliant.
  • Minimal extractables preserve fluid integrity.
  • Size: 1/16 in I.D., 3/16 in O.D., 50 ft coil.
  • Flexible with excellent fluid flow and easy to clean.