Learn More
Accuflow Systems Tygon Sanitary Silicone Tubing, 1/32 in ID, 3/32 in OD
Catalog No. NC2323786
Encompass_Preferred
Tygon SPT-3350 sanitary silicone tubing is a smooth-bore silicone tube designed for sanitary fluid transfer and food-and-beverage dispensing. The tubing provides an ultra-smooth inner surface to reduce particle entrapment and features low extractables to help preserve fluid integrity.
  • Ultra-smooth inner bore reduces particle entrapment.
  • Minimal extractables help maintain fluid integrity.
  • 1/32 in inner diameter, 3/32 in outer diameter, 1/32 in wall thickness.
  • Supplied on a 50 ft spool for continuous runs.
  • Wide recommended operating temperature range: -80 °C to 204 °C.
  • Suitable for sanitary food and beverage dispensing and general fluid transfer.
Catalog No. NC2323786 Supplier Accuflow Systems Supplier No. ABW00001
Tygon SPT-3350 sanitary silicone tubing is a smooth-bore silicone tube designed for sanitary fluid transfer and food-and-beverage dispensing. The tubing provides an ultra-smooth inner surface to reduce particle entrapment and features low extractables to help preserve fluid integrity.
  • Ultra-smooth inner bore reduces particle entrapment.
  • Minimal extractables help maintain fluid integrity.
  • 1/32 in inner diameter, 3/32 in outer diameter, 1/32 in wall thickness.
  • Supplied on a 50 ft spool for continuous runs.
  • Wide recommended operating temperature range: -80 °C to 204 °C.
  • Suitable for sanitary food and beverage dispensing and general fluid transfer.