Learn More
Accuflow Systems Tygon Silicone Tubing, 1/4 in ID, 50 ft
Catalog No. NC2323778
Encompass_Preferred
Sanitary silicone tubing with an ultra-smooth inner bore engineered for food and beverage dispensing and other sanitary fluid transfer applications. The smooth inner surface reduces particle entrapment and extractables, improves fluid flow, and facilitates cleaning and sanitization. Typical configuration is 1/4 in (0.250 in) inner diameter, 3/8 in (0.375 in) outer diameter, 1/16 in (0.063 in) wall thickness, 50 ft per pack.
  • Ultra-smooth inner bore reduces particle entrapment.
  • Minimal extractables help maintain fluid integrity.
  • Silicone construction offers flexibility and chemical compatibility.
  • 1/4 in ID, 3/8 in OD, 1/16 in wall, 50 ft length per pack.
  • Compliant with NSF-51 and FDA for food and beverage applications.
Catalog No. NC2323778 Supplier Accuflow Systems Supplier No. ABW00017
Sanitary silicone tubing with an ultra-smooth inner bore engineered for food and beverage dispensing and other sanitary fluid transfer applications. The smooth inner surface reduces particle entrapment and extractables, improves fluid flow, and facilitates cleaning and sanitization. Typical configuration is 1/4 in (0.250 in) inner diameter, 3/8 in (0.375 in) outer diameter, 1/16 in (0.063 in) wall thickness, 50 ft per pack.
  • Ultra-smooth inner bore reduces particle entrapment.
  • Minimal extractables help maintain fluid integrity.
  • Silicone construction offers flexibility and chemical compatibility.
  • 1/4 in ID, 3/8 in OD, 1/16 in wall, 50 ft length per pack.
  • Compliant with NSF-51 and FDA for food and beverage applications.