Learn More
Accuflow Systems Saint-Gobain Tygon SPT-3350 Tubing, 1/8-in ID, 1/4-in OD, 50 ft
Catalog No. NC2323780
Encompass_Preferred
Tygon SPT-3350 silicone tubing is a sanitary, platinum-cured silicone hose formulated for food, beverage, and other sanitary fluid transfer applications. Its smooth inner surface reduces particle entrapment and residue build-up, improving flow and facilitating effective cleaning and sanitation. Supplied in 50 ft rolls, the tube has a 1/8 in inner diameter, 1/4 in outer diameter, and a 1/16 in wall thickness.
  • Material: platinum-cured silicone.
  • Inner diameter: 1/8 in; outer diameter: 1/4 in; wall thickness: 1/16 in.
  • Packaged length: 50 ft per roll.
  • Meets FDA 21 CFR 175.300 and NSF 51.
  • Smooth inner surface reduces particle entrapment and improves cleanability.
  • Typical applications include food and beverage dispensing and sanitary fluid transfer.
Catalog No. NC2323780 Supplier Accuflow Systems Supplier No. ABW00007
Tygon SPT-3350 silicone tubing is a sanitary, platinum-cured silicone hose formulated for food, beverage, and other sanitary fluid transfer applications. Its smooth inner surface reduces particle entrapment and residue build-up, improving flow and facilitating effective cleaning and sanitation. Supplied in 50 ft rolls, the tube has a 1/8 in inner diameter, 1/4 in outer diameter, and a 1/16 in wall thickness.
  • Material: platinum-cured silicone.
  • Inner diameter: 1/8 in; outer diameter: 1/4 in; wall thickness: 1/16 in.
  • Packaged length: 50 ft per roll.
  • Meets FDA 21 CFR 175.300 and NSF 51.
  • Smooth inner surface reduces particle entrapment and improves cleanability.
  • Typical applications include food and beverage dispensing and sanitary fluid transfer.