Learn More
Accuflow Systems Tygon Silicone Tubing, 1/8-in ID x 3/16-in OD, Platinum-Cured
Catalog No. NC2323798
Encompass_Preferred
Platinum-cured sanitary silicone tubing engineered for food and beverage dispensing and sanitary fluid transfer. The ultra-smooth inner bore minimizes particle entrapment, improves fluid flow, and facilitates thorough cleaning and sanitization.
  • Platinum-cured silicone construction for sanitary applications.
  • Ultra-smooth inner bore reduces particle entrapment and extractables.
  • 1/8 in ID x 3/16 in OD, 0.031 in wall thickness.
  • Sold in 50 ft coils for process and laboratory use.
  • Operating temperature -80 °C to 204 °C (-112 °F to 400 °F).
  • Durometer Shore A 50 provides flexible, resilient performance.
  • Compliant with FDA material requirements and NSF-51 for food contact.
  • Suitable for sanitary fluid transfer, food and beverage dispensing, and cleaning-in-place systems.
Catalog No. NC2323798 Supplier Accuflow Systems Supplier No. ABW00006
Platinum-cured sanitary silicone tubing engineered for food and beverage dispensing and sanitary fluid transfer. The ultra-smooth inner bore minimizes particle entrapment, improves fluid flow, and facilitates thorough cleaning and sanitization.
  • Platinum-cured silicone construction for sanitary applications.
  • Ultra-smooth inner bore reduces particle entrapment and extractables.
  • 1/8 in ID x 3/16 in OD, 0.031 in wall thickness.
  • Sold in 50 ft coils for process and laboratory use.
  • Operating temperature -80 °C to 204 °C (-112 °F to 400 °F).
  • Durometer Shore A 50 provides flexible, resilient performance.
  • Compliant with FDA material requirements and NSF-51 for food contact.
  • Suitable for sanitary fluid transfer, food and beverage dispensing, and cleaning-in-place systems.