Learn More
Accuflow Systems Saint-Gobain Silicone Tubing, Sanitary, 3/8 in ID, 50 ft
Supplier:  Accuflow Systems ABW00028
Encompass_Preferred
Sanitary silicone tubing engineered for food and beverage dispensing and sanitary fluid transfer. The platinum-cured silicone formulation provides a smooth inner bore to minimize particle entrapment and residue while maintaining flexibility for processing and dispensing applications. Supplied in coils for easy installation in processing and dispensing lines.
  • Sanitary smooth inner surface to reduce residue and particle entrapment.
  • Platinum-cured silicone material for broad chemical compatibility and flexibility.
  • Dimensions: 3/8 in ID, 9/16 in OD, 3/32 in wall thickness.
  • 50 ft coil packaging for dispensing and transfer applications.
  • Suitable for food and beverage transfer and sanitary dispensing lines.
Catalog No. NC2323799