Learn More
Accuflow Systems Saint-Gobain Sanitary Silicone Tubing, 5/16 in ID, 1/2 in OD
Catalog No. NC2323812
Encompass_Preferred
Saint-Gobain Tygon SPT-3350 sanitary silicone tubing provides an ultra-smooth inner bore designed to reduce particle entrapment and simplify cleaning for hygienic food and beverage dispensing and sanitary fluid transfer.
  • Ultra-smooth inner bore reduces particle entrapment and eases sanitation.
  • Platinum-cured silicone for flexibility and thermal stability.
  • 5/16 in ID, 1/2 in OD, 3/32 in wall, 50 ft per pack.
  • Wide operating temperature range: -112 °F to 400 °F (-80 °C to 204 °C).
  • 50 Shore A durometer provides resilient, flexible performance.
  • Suitable for food and beverage dispensing and sanitary fluid transfer.
Catalog No. NC2323812 Supplier Accuflow Systems Supplier No. ABW00023
Saint-Gobain Tygon SPT-3350 sanitary silicone tubing provides an ultra-smooth inner bore designed to reduce particle entrapment and simplify cleaning for hygienic food and beverage dispensing and sanitary fluid transfer.
  • Ultra-smooth inner bore reduces particle entrapment and eases sanitation.
  • Platinum-cured silicone for flexibility and thermal stability.
  • 5/16 in ID, 1/2 in OD, 3/32 in wall, 50 ft per pack.
  • Wide operating temperature range: -112 °F to 400 °F (-80 °C to 204 °C).
  • 50 Shore A durometer provides resilient, flexible performance.
  • Suitable for food and beverage dispensing and sanitary fluid transfer.