Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, Platinum-Cured Silicone, 1/4 in ID, Braid-Reinforced
Catalog No. NC2323836
Encompass_Preferred
Braid-reinforced, platinum-cured silicone tubing with a 1/4 in (6.35 mm) inner diameter and 0.515 in (13.08 mm) outer diameter, supplied in 50 ft (15.24 m) coils. Designed for sanitary, high-purity fluid transfer and pressure applications in bioprocessing, pharmaceutical, and food and beverage manufacturing. Autoclavable and compatible with gas and radiation sterilization; engineered for elevated working pressure and thermal stability.
  • Braid-reinforced construction for elevated working pressure and durability.
  • Platinum-cured silicone provides chemical compatibility and thermal resistance.
  • 1/4 in (6.35 mm) I.D., 0.515 in (13.08 mm) O.D., 50 ft (15.24 m) coil.
  • Maximum working pressure 150 psi at 73°F, 105 psi at 320°F.
  • Autoclavable; compatible with ethylene oxide and gamma radiation sterilization.
  • Meets USP Class VI, FDA 21 CFR 177.2600, and 3-A sanitary standards.
Catalog No. NC2323836 Supplier Accuflow Systems Supplier No. AHJ001719
Braid-reinforced, platinum-cured silicone tubing with a 1/4 in (6.35 mm) inner diameter and 0.515 in (13.08 mm) outer diameter, supplied in 50 ft (15.24 m) coils. Designed for sanitary, high-purity fluid transfer and pressure applications in bioprocessing, pharmaceutical, and food and beverage manufacturing. Autoclavable and compatible with gas and radiation sterilization; engineered for elevated working pressure and thermal stability.
  • Braid-reinforced construction for elevated working pressure and durability.
  • Platinum-cured silicone provides chemical compatibility and thermal resistance.
  • 1/4 in (6.35 mm) I.D., 0.515 in (13.08 mm) O.D., 50 ft (15.24 m) coil.
  • Maximum working pressure 150 psi at 73°F, 105 psi at 320°F.
  • Autoclavable; compatible with ethylene oxide and gamma radiation sterilization.
  • Meets USP Class VI, FDA 21 CFR 177.2600, and 3-A sanitary standards.