Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuflow Systems Saint-Gobain Tygon Tubing, 5/16 in ID, 50 ft, Yellow

Catalog No. 502365881
Encompass_Preferred

Flexible tubing formulated to resist hydrocarbons and minimize swelling and embrittlement, with low extractables and translucent yellow appearance for flow monitoring.

  • Inside diameter 5/16 in, outside diameter 7/16 in, wall thickness 1/16 in.
  • Pack length 50 ft.
  • Color translucent yellow for visual identification and flow monitoring.
  • Formulation resists hydrocarbons and minimizes swelling and hardening.
  • Durometer hardness Shore A 57 for flexibility and durability.
  • Maximum recommended operating temperature 165°F (74°C); brittleness by impact -35°F (-37°C).
  • Suitable for fuel and lubricant applications including small engine fuel lines, coolant transfer, and general automotive use.

Catalog No. 50-236-5881 Supplier Accuflow Systems Supplier No. AAG00022
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flexible tubing formulated to resist hydrocarbons and minimize swelling and embrittlement, with low extractables and translucent yellow appearance for flow monitoring.

  • Inside diameter 5/16 in, outside diameter 7/16 in, wall thickness 1/16 in.
  • Pack length 50 ft.
  • Color translucent yellow for visual identification and flow monitoring.
  • Formulation resists hydrocarbons and minimizes swelling and hardening.
  • Durometer hardness Shore A 57 for flexibility and durability.
  • Maximum recommended operating temperature 165°F (74°C); brittleness by impact -35°F (-37°C).
  • Suitable for fuel and lubricant applications including small engine fuel lines, coolant transfer, and general automotive use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.